siRNA Details
Gene Symbol: Cpne8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025815.2 | NP_080091.1 | 12, 13 | 978 | ||
GenBank mRNA | AK004559.1 | - | 12, 13 | 966 | ||
BC017626.1 | - | 11, 12 | 872 | 4502073 | ||
BC138745.1 | - | 12, 13 | 977 | 8861767 | ||
BC138746.1 | - | 12, 13 | 977 | 8861768 |

More Information
Other Species Targeted

Gene Ontology Categories:
