siRNA Details
Gene Symbol: Pbrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081251.1 | NP_001074720.1 | 26 | 4140 | ||
GenBank mRNA | AK009120.1 | - | 2 | 166 | ||
BC055708.1 | - | 2 | 248 |

More Information
Other Species Targeted

Gene Ontology Categories:
