siRNA Details
Gene Symbol: Pbrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081251.1 | NP_001074720.1 | 28 | 4499 | ||
GenBank mRNA | AK009120.1 | - | 4 | 525 | ||
BC055708.1 | - | 4 | 607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
