siRNA Details
Gene Symbol: MTMR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011509099.2 | XP_011507401.1 | 14 | 1569 | ||
GenBank mRNA | AK296094.1 | - | 12 | 1395 | ||
BC094756.1 | - | 10 | 2750 | |||
U78556.1 | - | 12 | 1646 | |||
U78557.1 | - | 8 | 1051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
