siRNA Details
Gene Symbol: Kctd20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013254.1 | - | 3 | 1867 | ||
AK037247.1 | - | 1 | 1336 | |||
AK171742.1 | - | 7 | 2535 | |||
BC036318.1 | - | 6 | 2382 | |||
BC048161.1 | - | 7 | 2521 | |||
BC052712.1 | - | 7 | 2523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
