siRNA Details
Gene Symbol: 2010109I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025929.2 | NP_080205.1 | 3 | 212 | ||
GenBank mRNA | AK008365.1 | - | 3 | 211 | ||
AK018552.1 | - | 3 | 212 | |||
AK038164.1 | - | 188 | ||||
BC116397.1 | - | 3 | 195 | 40101946 | ||
BC116398.1 | - | 3 | 195 | 40101947 |
