siRNA Details
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027349.3 | NP_081625.3 | 15 | 2034 | ||
GenBank mRNA | AK011184.1 | - | 3 | 389 | ||
AK076553.1 | - | 1 | 162 | |||
AK087936.1 | - | 3 | 359 | |||
AK142132.1 | - | 9 | 1310 | |||
BC066150.1 | - | 4 | 468 | |||
BC067400.1 | - | 4 | 468 | |||
BC158104.1 | - | 14 | 1914 | 9007388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
