siRNA Details
Gene Symbol: D16Ertd472e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252438.1 | NP_001239367.1 | 5 | 2017 | ||
NM_001252439.1 | NP_001239368.1 | 5 | 1784 | |||
NM_001252440.1 | NP_001239369.1 | 5 | 1749 | |||
NM_025967.4 | NP_080243.3 | 5 | 1817 | |||
GenBank mRNA | AK009258.1 | - | 5 | 1988 | ||
BC019957.1 | - | 5 | 1708 | 3966397 |
