siRNA Details
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012645.1 | - | 4 | 800 | ||
AK013457.1 | - | 4 | 851 | |||
BC094292.1 | - | 4 | 797 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
