siRNA Details
Gene Symbol: Spag16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271533.1 | NP_001258462.1 | 4, 5 | 414 | ||
NM_029160.3 | NP_083436.2 | 4, 5 | 414 | |||
GenBank mRNA | AF490391.1 | - | 4, 5 | 396 | ||
AK015884.1 | - | 4, 5 | 415 | |||
AK083083.1 | - | 4, 5 | 393 | |||
AK162117.1 | - | 3, 4 | 329 | |||
BC120668.1 | - | 4, 5 | 413 | 40129591 | ||
BC145006.1 | - | 4, 5 | 413 | 9053554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
