siRNA Details
Gene Symbol: Spag16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271533.1 | NP_001258462.1 | 9, 10 | 898 | ||
NM_029160.3 | NP_083436.2 | 9, 10 | 898 | |||
GenBank mRNA | AF490391.1 | - | 9, 10 | 880 | ||
AK015884.1 | - | 9, 10 | 899 | |||
AK083083.1 | - | 9, 10 | 877 | |||
BC120668.1 | - | 9, 10 | 897 | 40129591 | ||
BC145006.1 | - | 9, 10 | 897 | 9053554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
