siRNA Details
Gene Symbol: Spag16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271533.1 | NP_001258462.1 | 7 | 697 | ||
NM_029160.3 | NP_083436.2 | 7 | 697 | |||
GenBank mRNA | AF490391.1 | - | 7 | 679 | ||
AK015884.1 | - | 7 | 698 | |||
AK083083.1 | - | 7 | 676 | |||
AK162117.1 | - | 6 | 612 | |||
BC120668.1 | - | 7 | 696 | 40129591 | ||
BC145006.1 | - | 7 | 696 | 9053554 |

More Information
Other Species Targeted

Gene Ontology Categories:
