siRNA Details
Gene Symbol: ZMYND11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK294469.1 | - | 12 | 1471 | ||
AK294781.1 | - | 13 | 1561 | |||
AK296427.1 | - | 12 | 1445 | |||
AK296518.1 | - | 9 | 1132 | |||
AK312570.1 | - | 14 | 1694 | |||
BC091489.1 | - | 13 | 1549 | 6186814 | ||
DQ335452.1 | - | 13 | 1616 | |||
DQ335453.1 | - | 12 | 1454 | |||
DQ335454.1 | - | 12 | 1454 | |||
DQ335455.1 | - | 13 | 1616 | |||
X86098.1 | - | 14 | 1740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
