siRNA Details
Gene Symbol: Nipbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027707.3 | NP_081983.2 | 47 | 9348 | ||
NM_201232.2 | NP_957684.1 | 46 | 9894 | |||
GenBank mRNA | AK020398.1 | - | 1 | 546 | ||
AK078435.1 | - | 1 | 664 | |||
AK135347.1 | - | 13 | 1450 | |||
BC049151.1 | - | 5 | 2420 | |||
BC055787.1 | - | 3 | 1201 |

More Information
Other Species Targeted

Gene Ontology Categories:
