siRNA Details
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013861.3 | NP_038889.1 | 9 | 1103 | ||
GenBank mRNA | AB027568.1 | - | 9 | 1104 | ||
AK028431.1 | - | 9 | 1126 | |||
AK033192.1 | - | 7 | 1207 | |||
AK034521.1 | - | 1112 | ||||
AK035044.1 | - | 9 | 1104 | |||
AK052225.1 | - | 9 | 1095 | |||
AK087844.1 | - | 9 | 1208 | |||
AK136486.1 | - | 8 | 957 | |||
AK195079.1 | - | 1 | 307 | |||
BC015246.1 | - | 9 | 1120 | 4221866 | ||
BC023354.1 | - | 8 | 932 | 5253545 |
