siRNA Details
Gene Symbol: Zfp292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013889.2 | NP_038917.2 | 8 | 4451 | ||
GenBank mRNA | AB093243.1 | - | 1 | 2023 | ||
AB221608.1 | - | 1 | 864 | |||
AF017806.1 | - | 1 | 2921 | |||
BC008537.1 | - | 1 | 539 |

More Information
Other Species Targeted

Gene Ontology Categories:
