siRNA Details
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015765.2 | NP_056580.2 | 12, 13 | 1512 | ||
GenBank mRNA | AK031489.1 | - | 1451 | |||
AK145937.1 | - | 12, 13 | 1466 | |||
AK153397.1 | - | 12, 13 | 1474 | |||
BC002056.1 | - | 12, 13 | 1440 | 3493119 | ||
BI734146.1 | - | 4, 5 | 615 | |||
U08215.1 | - | 12, 13 | 1478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
