siRNA Details
Gene Symbol: SRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006662.2 | NP_006653.2 | 16, 17 | 2886 | ||
GenBank mRNA | AB621816.1 | - | 16, 17 | 2953 | ||
AF143946.1 | - | 12, 13 | 2120 | |||
AK023808.1 | - | 9, 10 | 1599 | |||
AK128030.1 | - | 16, 17 | 2886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
