siRNA Details
Gene Symbol: Sall2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244916.1 | NP_001231845.1 | 2 | 397 | ||
NM_015772.3 | NP_056587.2 | 2 | 684 | |||
GenBank mRNA | AB093233.1 | - | 2 | 343 | ||
AJ007396.1 | - | 2 | 775 | |||
AK077601.1 | - | 2 | 311 | |||
BC054527.1 | - | 2 | 379 | |||
BC085361.1 | - | 2 | 461 | 5706710 | ||
CB520889.1 | - | 2 | 397 | |||
CB525158.1 | - | 2 | 466 |
