siRNA Details
Gene Symbol: Cfhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015780.2 | NP_056595.1 | 6 | 1018 | ||
GenBank mRNA | BC111870.1 | - | 6 | 1001 | 40057075 | |
M29008.1 | - | 6 | 1018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
