siRNA Details
Gene Symbol: Dlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194940.2 | NP_001181869.1 | 8, 9 | 2060 | ||
NM_001194941.1 | NP_001181870.1 | 4, 5 | 690 | |||
NM_015802.3 | NP_056617.2 | 4, 5 | 608 | |||
GenBank mRNA | AF178078.1 | - | 4, 5 | 544 | ||
AK147539.1 | - | 4, 5 | 690 | |||
AK162413.1 | - | 4, 5 | 609 | |||
AK166175.1 | - | 4, 5 | 568 | |||
BC150802.1 | - | 4, 5 | 393 | 9087713 | ||
GU169069.1 | - | 5, 6 | 314 | |||
HM008381.1 | - | 7, 8 | 1613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
