siRNA Details
Gene Symbol: Dlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194940.2 | NP_001181869.1 | 18 | 5246 | ||
NM_001194941.1 | NP_001181870.1 | 14 | 3876 | |||
NM_015802.3 | NP_056617.2 | 14 | 3794 | |||
GenBank mRNA | AF178078.1 | - | 14 | 3730 | ||
AK147539.1 | - | 14 | 3876 | |||
AK162413.1 | - | 14 | 3795 | |||
BC150802.1 | - | 14 | 3579 | 9087713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
