siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201569.1 | NP_001188498.1 | 5 | 716 | ||
NM_015805.3 | NP_056620.2 | 5 | 716 | |||
GenBank mRNA | AB206122.1 | - | 4 | 444 | ||
AF155913.1 | - | 4 | 444 | |||
AK030485.1 | - | 4 | 377 | |||
AK044331.1 | - | 5 | 691 | |||
AK166562.1 | - | 5 | 719 | |||
BC057000.1 | - | 5 | 641 | |||
BC079626.1 | - | 5 | 641 | 30536500 |
