siRNA Details
Gene Symbol: C1QL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006688.4 | NP_006679.1 | 2 | 923 | ||
GenBank mRNA | AF095154.1 | - | 2 | 708 | ||
BC008798.2 | - | 2 | 923 | 3635430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
