siRNA Details
Gene Symbol: Galns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193645.1 | NP_001180574.1 | 5 | 607 | ||
NM_016722.4 | NP_057931.3 | 5 | 607 | |||
GenBank mRNA | AK033946.1 | - | 5 | 525 | ||
AK159592.1 | - | 5 | 584 | |||
AK220245.1 | - | 5 | 509 | |||
BC004002.1 | - | 5 | 461 | 3492664 | ||
BQ550288.1 | - | 5 | 373 | |||
CB600418.1 | - | 2 | 68 |
