siRNA Details
Gene Symbol: MTMR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011509099.2 | XP_011507401.1 | 14 | 2738 | ||
GenBank mRNA | BC094756.1 | - | 10 | 3919 | ||
U78556.1 | - | 13 | 2365 | |||
U78557.1 | - | 9 | 1770 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
