siRNA Details
Gene Symbol: Tlr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016928.2 | NP_058624.2 | 4 | 654 | ||
GenBank mRNA | AF186107.1 | - | 1 | 1323 | ||
AK036842.1 | - | 4 | 656 | |||
BC125240.1 | - | 2 | 652 | 40043826 | ||
BC125247.1 | - | 2 | 652 | 40055024 | ||
BC125261.1 | - | 2 | 652 | 40048274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
