siRNA Details
Gene Symbol: MSMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006745.4 | NP_006736.1 | 2, 3 | 393 | ||
XM_005263176.2 | XP_005263233.1 | 2, 3 | 510 | |||
GenBank mRNA | AK292418.1 | - | 2, 3 | 338 | ||
AK309206.1 | - | 2, 3 | 338 | |||
BC010653.1 | - | 1, 2 | 287 | 3866053 | ||
BC107879.1 | - | 2, 3 | 355 | 6498540 | ||
BX441001.2 | - | 2, 3 | 393 | |||
U60205.1 | - | 1, 2 | 289 | |||
U93162.1 | - | 2, 3 | 353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
