siRNA Details
Gene Symbol: Copg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017477.2 | NP_059505.1 | 24 | 3593 | ||
GenBank mRNA | AF187079.1 | - | 24 | 3592 | ||
AK077461.1 | - | 24 | 3543 | |||
AK169256.1 | - | 24 | 3639 | |||
AK172519.1 | - | 24 | 3609 | |||
BC008553.1 | - | 16 | 2780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
