siRNA Details
Gene Symbol: IFRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006764.4 | NP_006755.4 | 1 | 89 | ||
GenBank mRNA | BI825477.1 | - | 1 | 89 | ||
Y12395.1 | - | 3 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
