siRNA Details
Gene Symbol: Zfp108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018791.2 | NP_061261.2 | 4 | 614 | ||
GenBank mRNA | AF167315.1 | - | 4 | 585 | ||
AK032355.1 | - | 4 | 615 | |||
BC100339.1 | - | 4 | 559 | 30939834 |
