siRNA Details
Gene Symbol: Zfp108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018791.2 | NP_061261.2 | 4 | 1632 | ||
GenBank mRNA | AF167315.1 | - | 4 | 1603 | ||
AK012570.1 | - | 406 | ||||
AK032355.1 | - | 4 | 1633 | |||
BC100339.1 | - | 4 | 1577 | 30939834 |
