siRNA Details
Gene Symbol: Cyp39a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285947.1 | NP_001272876.1 | 2 | 467 | ||
NM_001285948.1 | NP_001272877.1 | 2 | 467 | |||
NM_018887.4 | NP_061375.1 | 2 | 467 | |||
GenBank mRNA | AF237981.1 | - | 2 | 316 | ||
AK031733.1 | - | 2 | 415 | |||
BC021418.1 | - | 2 | 338 | 5025117 | ||
BC037663.1 | - | 2 | 316 | 5352740 |
