siRNA Details
Gene Symbol: Epb4.1l4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019427.2 | NP_062300.2 | 9 | 912 | ||
GenBank mRNA | AB032366.1 | - | 9 | 1407 | ||
AK144315.1 | - | 9 | 998 | |||
AK165783.1 | - | 9 | 905 | |||
BC113123.1 | - | 10 | 734 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001127565, BC166749.
Gene Ontology Categories:
