siRNA Details
Gene Symbol: Malat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020134.1 | - | 1 | 76 | ||
EF177380.1 | - | 1 | 150 | |||
FJ209304.1 | - | 1 | 150 |
Gene Symbol: Ramp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009681.1 | - | 152 | |||
AK019083.1 | - | 150 | ||||
AK028133.1 | - | 152 | ||||
AK084336.1 | - | 151 | ||||
AK088913.1 | - | 151 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC107410.1 | - | 96 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
