siRNA Details
Gene Symbol: UGT2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252274.2 | NP_001239203.2 | 2 | 212 | ||
NM_001252275.2 | NP_001239204.2 | 2 | 212 | |||
NM_001301239.1 | NP_001288168.1 | 1 | 150 | |||
NM_006798.4 | NP_006789.3 | 2 | 212 | |||
GenBank mRNA | AJ006054.1 | - | 1 | 159 | ||
AK304249.1 | - | 2 | 212 | |||
AK314209.1 | - | 2 | 210 | |||
BC142640.1 | - | 1 | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
