siRNA Details
Gene Symbol: Bace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019517.4 | NP_062390.3 | 6 | 1012 | ||
GenBank mRNA | AF051150.1 | - | 2 | 161 | ||
AF216310.1 | - | 6 | 963 | |||
AK052309.1 | - | 6 | 999 | |||
AK078770.1 | - | 6 | 979 | |||
AK165036.1 | - | 6 | 948 | |||
AK184226.1 | - | 3 | 252 | |||
AK185329.1 | - | 3 | 252 | |||
BC120773.1 | - | 6 | 978 | 40129696 | ||
BC131947.1 | - | 6 | 978 | 40130224 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001002802, XM_006248153, BC166597.
Gene Ontology Categories:
