siRNA Details
Gene Symbol: ERP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006817.3 | NP_006808.1 | 1, 2 | 270 | ||
GenBank mRNA | BC101493.1 | - | 1, 2 | 210 | 8068999 | |
BC101495.1 | - | 1, 2 | 210 | 8069001 | ||
CR541667.1 | - | 1, 2 | 152 | |||
FJ224317.1 | - | 1, 2 | 163 | |||
X94910.1 | - | 1, 2 | 163 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026129, AK009881, AK165702, BC017125, AB009293, and others.
Gene Ontology Categories:
