siRNA Details
Gene Symbol: Thsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205253.1 | NP_001192182.1 | 2 | 714 | ||
NM_001293688.1 | NP_001280617.1 | 3 | 857 | |||
NM_019576.2 | NP_062522.1 | 3 | 857 | |||
GenBank mRNA | AB039946.1 | - | 2 | 685 | ||
AK049636.1 | - | 3 | 820 | |||
AK052555.1 | - | 2 | 655 | |||
BC029898.1 | - | 3 | 847 | |||
BC051957.1 | - | 3 | 815 | |||
BC119603.2 | - | 2 | 685 | 40060210 |
