siRNA Details
Gene Symbol: Tspan6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019656.3 | NP_062630.2 | 5 | 613 | ||
GenBank mRNA | AF053454.1 | - | 5 | 560 | ||
AK146673.1 | - | 5 | 518 | |||
AK149204.1 | - | 5 | 523 | |||
AK160313.1 | - | 5 | 507 | |||
AK167384.1 | - | 5 | 509 | |||
BC003733.1 | - | 5 | 547 | 3590862 | ||
BC110314.1 | - | 5 | 522 | 5344878 |
