siRNA Details
Gene Symbol: ASCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284271.1 | NP_001271200.1 | 4 | 673 | ||
NM_006828.3 | NP_006819.2 | 4 | 673 | |||
XM_011535394.2 | XP_011533696.1 | 4 | 683 | |||
XM_011535395.2 | XP_011533697.1 | 5 | 908 | |||
XM_011535396.2 | XP_011533698.1 | 3 | 517 | |||
XM_017010205.1 | XP_016865694.1 | 5 | 929 | |||
XM_017010206.1 | XP_016865695.1 | 4 | 665 | |||
GenBank mRNA | AK299117.1 | - | 3 | 522 | ||
AK307903.1 | - | 4 | 602 | |||
AL834463.1 | - | 3 | 358 | |||
BC026066.1 | - | 4 | 670 | |||
CN255848.1 | - | 3 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
