siRNA Details
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019657.4 | NP_062631.1 | 11 | 1454 | ||
GenBank mRNA | AK012103.1 | - | 4 | 868 | ||
AK078841.1 | - | 11 | 1455 | |||
AK081869.1 | - | 11 | 1448 | |||
AK082715.1 | - | 11 | 1448 | |||
AK088521.1 | - | 11 | 1448 | |||
AK150849.1 | - | 11 | 1449 | |||
AK152489.1 | - | 11 | 1449 | |||
AK152835.1 | - | 1 | 392 | |||
AK166027.1 | - | 11 | 1448 | |||
BC090659.1 | - | 11 | 1424 | 30675153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
