siRNA Details
Gene Symbol: 1110004F10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019772.2 | NP_062746.1 | 5 | 697 | ||
GenBank mRNA | AB025407.1 | - | 5 | 697 | ||
AK003415.1 | - | 5 | 738 | |||
AK144059.1 | - | 5 | 724 | |||
AK167571.1 | - | 5 | 726 | |||
AK168206.1 | - | 5 | 731 | |||
AK169309.1 | - | 5 | 724 | |||
AK170172.1 | - | 3 | 455 | |||
AK187821.1 | - | 1 | 106 | |||
BC072575.1 | - | 5 | 733 | 5715413 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: FQ227306, BC089924.
Gene Ontology Categories:
