siRNA Details
Gene Symbol: Zmym3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177985.1 | NP_001171456.1 | 4 | 967 | ||
NM_001177986.1 | NP_001171457.1 | 4 | 967 | |||
NM_001177987.1 | NP_001171458.1 | 4 | 896 | |||
NM_001177988.1 | NP_001171459.1 | 4 | 814 | |||
NM_019831.3 | NP_062805.1 | 4 | 827 | |||
GenBank mRNA | AF156605.1 | - | 4 | 1453 | ||
AK033470.1 | - | 4 | 863 | |||
AK082075.1 | - | 4 | 810 | |||
AK122268.1 | - | 4 | 843 | |||
BC042699.1 | - | 4 | 832 | |||
BC050131.1 | - | 4 | 851 | |||
BC082320.1 | - | 4 | 896 | |||
BC108366.1 | - | 4 | 894 | |||
BC139001.1 | - | 3 | 760 | 8862023 | ||
BC145497.1 | - | 3 | 754 | 9054060 |

More Information
Other Species Targeted

Gene Ontology Categories:
