siRNA Details
Gene Symbol: Mpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164733.1 | NP_001158205.1 | 6 | 892 | ||
NM_001164734.1 | NP_001158206.1 | 6 | 902 | |||
NM_019939.2 | NP_064323.2 | 6 | 902 | |||
GenBank mRNA | AF161181.1 | - | 6 | 766 | ||
AF199009.1 | - | 6 | 898 | |||
AF199010.1 | - | 6 | 898 | |||
AK144434.1 | - | 6 | 903 | |||
AK164549.1 | - | 6 | 893 | |||
BC042868.2 | - | 6 | 849 | |||
BC046421.1 | - | 6 | 849 | |||
BC069908.1 | - | 6 | 891 | |||
BC106140.1 | - | 6 | 849 | |||
BC138666.1 | - | 6 | 760 | 8861687 | ||
BC145363.1 | - | 6 | 760 | 9053921 | ||
BC145364.1 | - | 6 | 750 | 9053922 |
