siRNA Details
Gene Symbol: INMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199219.1 | NP_001186148.1 | 2 | 214 | ||
NM_006774.4 | NP_006765.4 | 2 | 217 | |||
GenBank mRNA | AF128846.1 | - | 2 | 217 | ||
AF128847.1 | - | 2 | 217 | |||
AK309519.1 | - | 3 | 347 | |||
AK313832.1 | - | 2 | 212 | |||
BC033813.1 | - | 2 | 199 | |||
BC103712.1 | - | 2 | 200 | |||
BC106902.1 | - | 2 | 217 | 40031569 | ||
BC106903.1 | - | 2 | 217 | 40031570 | ||
DA920005.1 | - | 2 | 217 |
Gene Symbol: INMT-FAM188B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037598.1 | 2 | 214 | |||
GenBank mRNA | AK308762.1 | - | 2 | 212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
