siRNA Details
Gene Symbol: Zfp109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020262.3 | NP_064658.1 | 4 | 738 | ||
GenBank mRNA | AF167316.1 | - | 4 | 700 | ||
BC119619.1 | - | 3 | 307 | 40062160 | ||
BC119620.1 | - | 4 | 365 | 40062167 |
