siRNA Details
Gene Symbol: C6orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286474.1 | NP_001273403.1 | 10, 11 | 506 | ||
NM_001286475.1 | NP_001273404.1 | 9, 10 | 485 | |||
GenBank mRNA | AK225742.1 | - | 10, 11 | 497 | ||
AK302176.1 | - | 9, 10 | 445 | |||
AK310230.1 | - | 9, 10 | 474 | |||
DC399589.1 | - | 10, 11 | 507 | |||
U60665.1 | - | 10, 11 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
