siRNA Details
Gene Symbol: Angel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199020.1 | NP_001185949.1 | 10 | 2687 | ||
NM_021421.4 | NP_067396.3 | 9 | 2822 | |||
GenBank mRNA | AK077039.1 | - | 11 | 2817 | ||
AK088834.1 | - | 8 | 2454 | |||
AK163397.1 | - | 1 | 865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
