siRNA Details
Gene Symbol: Nrbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036293.2 | NP_001031370.1 | 4 | 1198 | ||
GenBank mRNA | AK002810.1 | - | 4 | 1197 | ||
AK004200.1 | - | 4 | 1118 | |||
BC019448.1 | - | 4 | 1198 | 4212132 |

More Information
Other Species Targeted

Gene Ontology Categories:
